missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Tartaric acid, ACS, Thermo Scientific Chemicals

Useful as a resolving agent

43.11 € - 361.80 €

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
SMILES OC(C(O)C(O)=O)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11377868
View Documents
Thermo Scientific Alfa Aesar
036405.22
100 g
43.11 €
100g
Please to purchase this item. Need a web account? Register with us today!
11397868
View Documents
Thermo Scientific Alfa Aesar
036405.36
500 g
120.60 €
500g
Please to purchase this item. Need a web account? Register with us today!
11387868
View Documents
Thermo Scientific Alfa Aesar
036405.A3
2 kg
361.80 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

L-(+)-Tartaric acid is widely utilized in pharmaceutical industries. It is used in soft drinks, confectionaries, food products, gelatin desserts and buffering agents. It forms a compound, TiCl2(O-i-Pr)2 with Diels-Alder catalyst and act as a chelate agent in metal industries. Owing to its efficient chelating property towards metal ions, it is used in the farming and metal industries for complexing micronutrients and for cleaning metal surfaces, respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

87-69-4
150.09
FEWJPZIEWOKRBE-UHFFFAOYNA-N
444305
OC(C(O)C(O)=O)C(O)=O
C4H6O6
MFCD00064207
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
CHEBI:15671

Specifications

87-69-4
C4H6O6
1725147
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
FEWJPZIEWOKRBE-UHFFFAOYNA-N
2,3-dihydroxybutanedioic acid
444305
150.09
1.76
L-(+)-Tartaric Acid
168°C to 170°C
MFCD00064207
14,9070
Soluble in water,methanol and acetone.
OC(C(O)C(O)=O)C(O)=O
150.09
CHEBI:15671
ACS
Granular
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318

P280-P305+P351+P338-P310

H318-H500

missing translation for 'einecsNumber' : 201-766-0

missing translation for 'rtecsNumber' : WW7875000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.