missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Tartaric Acid, Granular, Multi-Compendial, N.F., J.T. Baker™

Product Code. 15538254
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 87-69-4
Molecular Formula C4H6O6
MDL Number MFCD00064207
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILES OC(C(O)C(O)=O)C(O)=O
IUPAC Name 2,3-dihydroxybutanedioic acid
Molecular Weight (g/mol) 150.09
PubChem CID 444305
ChEBI CHEBI:15671
Formula Weight 150.09
Grade Multi-Compendial/NF
Quantity 2.5 kg
Packaging Poly Bottle
Physical Form Granular
Specific Gravity 1L = 1.76kg
Chemical Name or Material L-(+)-Tartaric Acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'