missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Threonine, 98% Product Code.: 10665762

Thermo Scientific Chemicals L-Threonine, 98%

Product Code. 10665762
25 g, Glass Bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10665762

Brand: Thermo Scientific Chemicals 138930250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-19-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064270
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
Synonym l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
PubChem CID 6288
ChEBI CHEBI:16857
SMILES CC(O)C(N)C(O)=O

Specifications

Melting Point 255.0°C
Color Yellow
Chloride 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Loss on Drying 0.5 % max. (1 g, 105°C, 3 hrs)
Packaging Glass Bottle
Linear Formula CH3CH(OH)CH(NH2)CO2H
Specific Rotation − 28.30
Sulfate 300ppm max.
Merck Index 15, 9534
Sulfated Ash 0.1 % max.
Solubility Information Solubility in water: 90g/L (20°C). Other solubilities: insoluble in most common organic solvents
IUPAC Name (2S,3R)-2-amino-3-hydroxybutanoic acid
Specific Rotation Condition −28.30 (20.00°C c=6,H2O)
Quantity 25 g
Formula Weight 119.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L-Threonine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466042 Certificate TypeCertificate of Analysis Date16/09/2024 Catalog Number138930050,138931000,138930250,138935000
Lot NumberA0429075 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog Number138930050,138931000,138930250,138935000
Lot NumberA0460420 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number138930050,138931000,138930250,138935000
Lot NumberA0442828 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog Number138930050,138931000,138930250,138935000
Lot NumberA0451303 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number138930050,138931000,138930250,138935000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Threonine, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.