missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Threonine, Cell Culture Reagent

Product Code. 15405899
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
1 kg
missing translation for 'unitSize'
1kg
250g
This item is not returnable. View return policy

Product Code. 15405899

missing translation for 'mfr': Thermo Scientific Chemicals J63709.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72-19-5 | C4H9NO3 | 119.12 g/mol

Threonine is used to treat various nervous system disorders including spinal spasticity, multiple sclerosis, familial spastic paraparesis, and amyotrophic lateral sclerosis (ALS, Lou Gehrig's disease). Useful with intestinal disorders and indigestion, threonine has also been used to alleviate anxiety and mild depression. . L-threonine has been used clinically with the aim of increasing glycine concentrations in the cerebral spinal fluid of patients with spasticity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water 50 mg/mL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-19-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064270
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
Synonym l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
PubChem CID 6288
ChEBI CHEBI:16857
IUPAC Name (2S,3R)-2-amino-3-hydroxybutanoic acid
SMILES CC(O)C(N)C(O)=O

Specifications

Melting Point 256°C (decomposition)
Color White
Beilstein 1721646
Merck Index 14,9380
Solubility Information Soluble in water 50mg/mL.
Quantity 1 kg
Formula Weight 119.12
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Threonine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.