missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Thyroxine T4 Monoclonal Antibody (T4YCH), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514716
This item is not returnable.
View return policy
Description
MA5-14716 targets L-Thyroxine (T4) in ELISA, Western Blot, IP, and RIA applications and shows reactivity with Chemical samples. Product MA514716 is a smaller package size of MIT0501 (formerly sold as a Seradyn product).
Levothyroxine, also L-thyroxine, synthetic T4, or 3,5,3',5'-tetraiodo-L-thyronine, is a synthetic form of thyroxine (thyroid hormone), used as a hormone replacement for patients with thyroid problems. The natural hormone is chemically in the chiral L-form, as is the pharmaceutical agent. Dextrothyroxine (D-thyroxine) briefly saw research as an anticholesterol agent but was pulled due to cardiac side-effects.Specifications
L-Thyroxine T4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG2b |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
T4YCH | |
Unconjugated | |
C15H11I4NO4; Levothyroxine; L-thyroxine; Synthroid; Thyroxine | |
Thyroxine-BSA | |
100 μg | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid | |
Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction