missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Tryptophan, Cell Culture Reagent Product Code.: 15414889

Thermo Scientific Chemicals L-Tryptophan, Cell Culture Reagent

Product Code. 15414889
10 g
Click to view available options
Quantity:
10 g
100 g
1 kg
This item is not returnable. View return policy

Product Code. 15414889

Brand: Thermo Scientific Chemicals J62508.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-22-3
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064340
InChI Key QIVBCDIJIAJPQS-VIFPVBQESA-N
Synonym l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
PubChem CID 6305
ChEBI CHEBI:16828
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point 280°C to 285°C
Color White
Beilstein 86197
Merck Index 14,9797
Quantity 10 g
Formula Weight 204.23
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Tryptophan

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Tryptophan, Cell Culture Reagent >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.