Learn More
Thermo Scientific Chemicals L-Tyrosine disodium salt dihydrate, Thermo Scientific Chemicals
Description
Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
Solubility
Soluble in water.
Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Color | Brown to White |
Solubility Information | Soluble in water. |
IUPAC Name | 2-amino-3-(4-hydroxyphenyl)propanoic acid dihydrate disodium |
Quantity | 1 kg |
Formula Weight | 261.18 (225.15 Anhydrous) |
Optical Rotation | −13° (c=4 in Water) |
Sensitivity | Very Hygroscopic |
Percent Purity | 99% |
Physical Form | Powder |
Chemical Name or Material | L-Tyrosine disodium salt dihydrate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y04K056 | Certificate of Analysis | 14/11/2024 | J61770.22,J61770.A1,J61770.36 |
Z07J028 | Certificate of Analysis | 09/01/2024 | J61770.22,J61770.A1,J61770.36 |
T11I009 | Certificate of Analysis | 22/07/2022 | J61770.22,J61770.A1,J61770.36 |
Z10F052 | Certificate of Analysis | 08/12/2021 | J61770.22,J61770.A1,J61770.36 |
F29X159 | Certificate of Analysis | 08/12/2021 | J61770.22,J61770.A1,J61770.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.