missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Valine, 99% Product Code.: 11460653

Thermo Scientific Chemicals L-Valine, 99%

Product Code. 11460653
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11460653

Brand: Thermo Scientific Chemicals A12720.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Valine is used in the enhancement of muscle growth and energy. It is utilized in tissue repair. It is also involved in the studies to attenuate arrhythmias and induce hypotensive effects. It acts as a precursor for the preparation of penicillin through biosynthetic pathway. It also employed as nutritional supplements in food and beverage industries. Further, it plays an important role in maintaining mental vigor, muscle coordination and emotional calm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. Slightly soluble in methanol and acetone.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-18-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064220
InChI Key KZSNJWFQEVHDMF-BYPYZUCNSA-N
Synonym l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin
PubChem CID 6287
ChEBI CHEBI:16414
IUPAC Name (2S)-2-amino-3-methylbutanoic acid
SMILES CC(C)[C@H](N)C(O)=O

Specifications

Melting Point ∽315°C
Assay Percent Range 99%
Beilstein 1721136
Merck Index 14,9909
Solubility Information Soluble in water. Slightly soluble in methanol and acetone.
Density 1.23
Quantity 25 g
Formula Weight 117.15
Percent Purity 99%
Chemical Name or Material L-Valine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Valine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.