missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Valine methyl ester hydrochloride, 99% Product Code.: 10318320

Thermo Scientific Chemicals L-Valine methyl ester hydrochloride, 99%

Product Code. 10318320
10 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10318320

Brand: Thermo Scientific Chemicals 229260100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6306-52-1
Molecular Formula C6H13ClNO2
Molecular Weight (g/mol) 166.63
MDL Number MFCD00012497
InChI Key RFMIEHOYULVMNZ-UHFFFAOYNA-N
Synonym l-valine methyl ester hydrochloride, h-val-ome.hcl, methyl valinate hydrochloride, s-methyl 2-amino-3-methylbutanoate hydrochloride, h-l-val-ome hcl, methyl l-valinate hydrochloride, valine methyl ester hydrochloride, s-2-amino-3-methyl-butyric acid methyl ester hydrochloride, h-val-ome hcl, methyl 2s-2-amino-3-methylbutanoate hydrochlorideShow MoreShow Less
PubChem CID 111190
IUPAC Name methyl (2S)-2-amino-3-methylbutanoate;hydrochloride
SMILES [Cl].COC(=O)C(N)C(C)C

Specifications

Melting Point 171.0°C to 173.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)2CHCH(NH2)COOCH3·HCl
Quantity 10 g
Specific Rotation Condition +15.50° (24°C C=2,H2O)
Specific Rotation + 15.50
Solubility Information (5% in water) Clear colorless to light yellow
Formula Weight 167.64
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L-Valine methyl ester hydrochloride, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461842 Certificate TypeCertificate of Analysis Date09/04/2024 Catalog Number229260100,229260500
Lot NumberA0458092 Certificate TypeCertificate of Analysis Date27/11/2023 Catalog Number229260100,229260500
Lot NumberA0380482 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number229260100,229260500
Lot NumberA0403534 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog Number229260100,229260500
Product Identifier
  • L-Valine methyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Valine methyl ester hydrochloride, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.