Learn More
Epredia™ Lab Vision™ CD138 Ab-2, Mouse Monoclonal Antibody
Obtain accurate and reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.
144.00 € - 682.00 €
Specifications
Antigen | CD138 Ab-2 |
---|---|
Clone | MI15 |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12660157
|
Epredia™
MS-1793-S0 |
Concentrated | 100 μL | N/A | |||||
12670157
|
Epredia™
MS-1793-S1 |
Concentrated | 500 μL |
455.00 €
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12650157
|
Epredia™
MS-1793-S |
Concentrated | 1 mL |
682.00 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12630157
|
Epredia™
MS-1793-R7 |
Prediluted | 7 mL |
294.00 €
7mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12640157
|
Epredia™
MS-1793-RQ |
Ready-to-Use | 12 mL |
434.00 €
12mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CD138/Syndecan-1 is a transmembrane heparin sulphate proteoglycan which is made up of one core protein and five glycosaminoglycan. CD138 is expected to play a role in cell adhesion. It is expressed on the surface of pre-B cells and plasma cells but is absent from mature B cells. It is a selective marker for B cell lymphoblastic leukemia and lymphoplasmocytoid leukemia. It is lost from the apoptotic myeloma cells; hence, it is a useful marker for viable myeloma cells.Host Species: Mouse
Clone: MI15
Isotype: IgG1/κ
Species Reactivity: Human. Others not known
Epitope: Ectodomain
Immunogen: U-266 cells
Molecular Weight: 30.5kDa (core protein); 85-92kDa (depending upon glycosaminoglycans)
Positive Control: U-266 cells, Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Specifications
CD138 Ab-2 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
IVD | |
U-266 cells |
MI15 | |
Monoclonal | |
Mouse | |
Cell Adhesion | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.