missing translation for 'onlineSavingsMsg'
Learn More

CD5 Rabbit Monoclonal Antibody, Epredia™

Obtain accurate and reproducible results in immunohistochemistry and western blotting procedures with Epredia™ CD5, Rabbit Monoclonal Antibody.

Brand:  Epredia RM-9119-RQ

 View more versions of this product

Product Code. 12673757

  • 708.00 € / 12mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CD5, a transmembrane protein, is found on 95%of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Occasionally, CD5 antigen is also expressed on a subset of B cells. Mantle cell lymphomas (same as diffuse centrocytic lymphomas) are CD5+ while the follicle center cell lymphoma are CD5-.

Host Species: Rabbit

Clone: SP19

Species Reactivity: Human. Others not tested.

Epitope: Intracellular region

Immunogen: Synthetic peptide from the intracellular region of CD5

Positive Control: Raji cells, Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Specifications

Specifications

CD5
Monoclonal
Unconjugated
Synthetic peptide from the intracellular region of CD5
IVD
Primary
Western Blot
SP19
Rabbit
12 mL
Hematopoietic
Ready-to-Use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.