missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD8 Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD8 Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-457-S1

 View more versions of this product

Product Code. 12631497

  • 703.00 € / 500µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CD8 molecule consists of two chains, termed α and Β chain, which are expressed as a disulphide-α β heterdimer or as an alpha α homodimer on T cell subset (cytotoxic/suppressor T cells), thymocytes and NK cells. The majority of CD8+ Tcells expresses CD8 as αβheterodimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 α chain (but not Β chain).

Host Species: Mouse

Clone: C8/144B

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: C-terminal

Immunogen: A 13 amino acid synthetic peptide from the C-terminal cytoplasmic domain of α-chain of human CD8 molecule

Molecular Weight: 32 kDa

Positive Control: Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD8 Ab-1
Monoclonal
Unconjugated
A 13 amino acid synthetic peptide from the C-terminal cytoplasmic domain of alpha chain of human CD8 molecule
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
C8/144B
Mouse
500 μL
T-Cell
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.