missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ EDTA Buffer for Heat-Induced Epitope Retrieval (10X)

Improve the reactivity of many antibodies in formalin-fixed tissues with Epredia™ Lab Vision™ EDTA Buffer for Heat-Induced Epitope Retrieval (10X).

183.00 €

Specifications

Content And Storage Room temperature
Dilution 10X
Product Type EDTA
For Use With (Application) Immunohistochemistry
pH 8
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12668286
View Documents
Epredia™
AP-9004-500
500mL N/A N/A N/A
Description
Show More
Specifications

Specifications

Room temperature
EDTA
8
10X
Immunohistochemistry
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ Lab Vision™ EDTA Buffer for Heat-Induced Epitope Retrieval (10X)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.