Learn More
Epidermal Growth Factor Receptor/EGFR Ab-11 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments.
141.78 € - 700.00 €
Specifications
Antigen | Epidermal Growth Factor Receptor/EGFR Ab-11 |
---|---|
Clone | 199.12 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Concentrated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12661307
|
Epredia
MS-396-P0 |
200μg/mL | 100 μL | N/A | |||||
12691307
|
Epredia
MS-396-P1 |
200μg/mL | 500 μL |
459.00 €
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12671307
|
Epredia
MS-396-P |
200μg/mL | 1 mL | N/A | |||||
12611317
|
Epredia
MS-396-P1ABX |
1mg/mL | 100 μL |
504.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12631317
|
Epredia
MS-396-PABX |
1mg/mL | 200 μL | N/A | |||||
Description
EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.Host Species: Mouse
Clone: 199.12
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Epitope: Extracellular domain
Immunogen: Extracellular domain of human recombinant EGFR protein
Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)
Positive Control: A431 cells
Cellular Localization: Membrane
Recommended for:
- Western Blotting
- Immunoprecipitation (Native only)
- Effect on EGF Binding (Noninhibitory)
Note:
USA: RUO; Int'l: RUO
Specifications
Epidermal Growth Factor Receptor/EGFR Ab-11 | |
Monoclonal | |
Concentrated | |
RUO | |
Human | |
IgG2a |
199.12 | |
Unconjugated | |
Mouse | |
EGFR Family | |
Extracellular domain of human recombinant EGFR protein | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.