missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ HIER Buffer H (100X)

Perform heat induced epitope retrieval (HIER) on formalin-fixed paraffin-embedded tissue sections using the Epredia™ Lab Vision™ HIER Buffer H (100X).

Brand:  Epredia™ TA-135-HBH


Product Code. 12603987

  • 662.00 € / 135mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • For in vitro diagnostic use
  • Store at room temperature.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Room temperature
HIER
Immunohistochemistry
IVD
100X
9 x 15 mL
6 to 7
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : EDTA, tetrasodium EDTA, 2-methyl-3-isothiazolone, tris (hydroxymethyl) aminomethane, triton-X 100

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ Lab Vision™ HIER Buffer H (100X) > 9 x 15mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.