missing translation for 'onlineSavingsMsg'
Learn More

pan-Actin Ab-5 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry, western blotting, and immunoprecipitation experiments.

Brand:  Epredia MS-1295-P1ABX

 View more versions of this product

Product Code. 12659496

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Host Species: Mouse

Clone: ACTN05; same as C4

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Mouse, Dog, Pig, Rabbit, Rat, Chicken, Dictyostelium discoideum and Physarum polycephalum. Others not known.

Epitope: Not determined

Immunogen: Chicken gizzard actin

Positive Control: HeLa cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Actin, pan Ab-5
Monoclonal
1mg/mL
Mouse
100 μL
Muscle Biology
Bovine, Canine, Chicken, Human, Mouse, P. polycephalum, Porcine, Rabbit, Rat
IgG1 κ
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
ACTN05; same as C4
Unconjugated
Chicken gizzard actin
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.