Learn More
Epredia™ Lab Vision™ Placental Alk. Phosphatase (PLAP) Ab-4, Mouse Monoclonal Antibody
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ Placental Alk. Phosphatase (PLAP) Ab-4, Mouse Monoclonal Antibody.
124.44 € - 495.00 €
Specifications
Antigen | Placental Alk. Phosphatase (PLAP) Ab-4 |
---|---|
Clone | 8A9 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12642197
|
Epredia™
MS-734-S0 |
Concentrated | 100 μL | N/A | |||||
12652197
|
Epredia™
MS-734-S1 |
Concentrated | 500 μL |
362.00 €
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12632197
|
Epredia™
MS-734-S |
Concentrated | 1 mL |
495.00 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12622197
|
Epredia™
MS-734-R7 |
Prediluted | 7 mL | N/A | |||||
Description
Host Species: Mouse
Clone: 8A9
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Purified human Placental Alkaline Phosphatase (PLAP)
Molecular Weight: 60-70kDa
Positive Control: Placenta or seminoma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Placental Alk. Phosphatase (PLAP) Ab-4 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgG2a κ |
8A9 | |
Monoclonal | |
Mouse | |
Cancer and Tumor Biology | |
Purified human Placental Alkaline Phosphatase (PLAP) | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.