missing translation for 'onlineSavingsMsg'
Learn More

Prohibitin (Mitochondrial Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Prohibitin (Mitochondrial Marker) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-261-P1ABX

 View more versions of this product

Product Code. 12690727

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Prohibitin is an evolutionarily conserved protein located in the inner membrane of mitochondria. Prohibitin appears to be a very reliable marker of mitochondria. Prohibitin shows antiproliferative activity and has been proposed to play a role in normal cell cycle regulation, replicative senescence, cellular immortalization, and tumor suppression.

Host Species: Mouse

Clone: II-14-10

Isotype: IgG1

Species Reactivity: Human, Pig, Rat, Mouse, and Chicken. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant rat prohibitin protein

Molecular Weight: 30kDa

Positive Control: MAD109 or PC12 cells, Heart

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Prohibitin (Mitochondrial Marker) Ab-1
Monoclonal
1mg/mL
Mouse
100 μL
Apoptosis
Chicken, Human, Mouse, Pig, Rat
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
II-14-10
Unconjugated
Purified recombinant rat prohibitin protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.