Learn More
Epredia™ Lab Vision™ PT Module™ Deparaffinization and Heat-Induced Epitope Retrieval Solutions (100X), Citrate buffer, pH 6
Perform Heat Induced Antigen Retrieval (HIER) on formalin fixed paraffin-embedded tissue sections with Epredia™ Lab Vision™ PT Module™ Epitope Retrieval Solutions.
Brand: Epredia™ TA-125-PM2X
This item is not returnable.
View return policy
Description
- 100X stock solution
- Dilute 100-fold with distilled water before use
- Designed for use with Pretreatment Module™
- Available in Citrate, EDTA or Tris-HCl at various pH levels
Storage Conditions: Store at room temperature. Stable for up to 18 months. Contains no preservatives; for periods longer than three months, store at 2°-8°C.
Specifications
Room temperature | |
HIER | |
Immunohistochemistry |
100X | |
125mL | |
8 |
Product Content Correction
Product Title
Epredia™ Lab Vision™ PT Module™ Deparaffinization and Heat-Induced Epitope Retrieval Solutions (100X), Citrate buffer, pH 6 > EDTA buffer, pH 8; 125mL
Spot an opportunity for improvement?Share a Content Correction