missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ UltraVision™ ONE Detection System: HRP Polymer (Ready-To-Use)

A one-step polymer system providing increased sensitivity, time savings and detection simplicity.

Brand:  Epredia™ TL-125-HLJ

 View more versions of this product

Product Code. 12644017

  • 2165.00 € / 125mL
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

UltraVision ONE is a robust ONE-step polymer system that provides increased sensitivity, time savings and detection simplicity. The UltraVision ONE HRP polymer is an innovative, patented technology. It consists of smaller amino acid polymer subunits that minimize conflicts in binding the target protein. Decreased binding conflicts result in more consistent staining and better signal amplification. Ultimately, this gives the user higher sensitivity and antibody efficiency. UltraVision ONE HRP polymer allows the use of less antibody to obtain better signal-to-noise ratios. This system is also biotin-free, which eliminates background staining found with traditional biotin-based detection methods. For optimal interpretation of results, appropriate positive and negative controls must be included.
  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Peroxidase

  • Chromogen/Substrate: None provided

Includes:

TL-060-HL: 60mL Ultra V Block (TA-060-UB), 60mL UltraVision ONE HRP Polymer (TL-060-PHJ)

TL-125-HL: 125mL Ultra V Block (TA-125-UB), 125mL UltraVision ONE HRP Polymer (TL-125-PHJ)

Note:

UltraVision ONE HRP polymer detection system detects rabbit and mouse immunoglobulins bound to an antigen in tissue sections. The specific primary antibody is located by a universal secondary antibody polymer formulation. The amino acid polymer is conjugated to horseradish peroxidase and the Fab fragments of goat anti-rabbit and goat anti-mouse. The polymer complex is then visualized with an appropriate substrate/chromogen.

Specifications

Specifications

625 to 1250 tests
125mL Ultra V Block (TA-125-UB), 125mL UltraVision ONE HRP Polymer (TL-125-PHJ)
anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Peroxidase
None provided
125 mL
None provided
Store at 2° to 8°C. Each component is stable for 18 months.
Lab Vision™, UltraVision™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.