missing translation for 'onlineSavingsMsg'
Learn More

Vimentin Ab-2 Mouse Monoclonal Antibody, Epredia™

Epredia™ Vimentin Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-129-PABX

 View more versions of this product

Product Code. 12639516

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Vimentin is the main intermediate filament protein in mesenchymal cells and is therefore of value in the differential diagnosis of undifferentiated neoplasms.

Host Species: Mouse

Clone: V9

Isotype: IgG1/κ

Species Reactivity: Rat, Horse, Chicken, Cow, Cat, Dog, Human and Pig. Others not known.

Epitope: Not determined

Immunogen: Purified vimentin from pig eye lens

Molecular Weight: 57-60kDa

Positive Control: Sarcomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Vimentin Ab-2
Monoclonal
1mg/mL
Mouse
200 μL
Cytoskeletal Protein
Bovine, Canine, Chicken, Equine, Feline, Gerbil, Hamster, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat
IgG2a κ
Immunohistochemistry (Paraffin)
V9
Unconjugated
Purified vimentin from pig eye lens
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.