Learn More
Labconco™ California Büchner Funnel
For use with Labconco Crude Fiber Apparatus.
Brand:  Labconco™ 5510000
Product Code. 16211372
  • 204.00 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Stainless steel and polyethylene, two piece Buchner Funnel is modified by the addition of a 200-mesh stainless steel screen that is firmly heat sealed into the bottom of the upper section.
Specifications
Specifications
Polyethylene
Crude fiber apparatus