missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ CentriVap™ Concentration Systems: Ultra-Low (-84°C) Cold Trap

For use with Labconco CentriVap Benchtop Concentrator

Brand:  Labconco™ 7460030

 View more versions of this product

Product Code. 10523693

  • 8765.00 € / Each
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Protect vacuum pump by trapping moisture, vapors and corrosive fumes as they evaporate from samples
  • Environment-friendly, CFC-free refrigerant
  • Stainless-steel condenser collects 4L of liquid, accepts optional glass insert (16-315-61) for corrosives
  • Corrosion-resistant drain speeds cleanup
  • READY light illuminates at -40°C, indicating concentration conditions are right

Order Info

Available in 115V and 230V models.

Compliance

ETL, ETL-C, and CE listed

Specifications

Specifications

CE, ETL, ETL-C
Low freezing point solvent concentrator system
Stainless Steel
NEMA 6-15
22 in.
36.8 cm
53.1 cm
55.9 cm
64 kg
230 V
50 Hz
-84°C
1/3hp Compressor, 4L Stainless Steel collector, Acrylic lid with two quick-disconnect fittings, Stainless Steel drain port with Nylon valve, grounding clip and on/off power switch
-65°C
Benchtop
14.5 in.
20.9 in.
22 in.
142 lb.
230 V 50 Hz
6 A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.