Learn More
Labconco™ Drying Chamber Valves for FreeZone™ Freeze-Dry Systems
Replacement valves for drying chamber or manifold ports provide leak-resisting high-vacuum seals.
Brand: Labconco™ 7509600
Requires : Ampule adapters are available for connecting ampules to 1/2 in. valve.
This item is not returnable. View return policy
Description
Replacement valves for drying chamber or manifold ports provide leak-resisting high-vacuum seals.
- Fabricated of neoprene body and gaskets, molded plastic knob and fittings
- Valve 10-273-12 accommodates both 1/2 and 3/4 in. adapters
- Serum bottles fit in valves without adapter
†Drying chambers, manifolds, and support shelves for stoppering tray dryer.
Order Info
Contact your Customer Service Representative for assistance in selecting the proper valve.
Specifications
Drying Chamber Valve | |
Drying chambers and manifolds | |
3 in. | |
2 in. | |
0.5 kg |
Neoprene | |
Freeze-Dry System | |
7.62 cm | |
1 lb. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.