Learn More
Labconco™ Fast-Freeze™ Flask Adapters
For connecting Fast-Freeze™ flasks to freeze-dryer valves
Brand:  Labconco™ 7545200
 View more versions of this product
Product Code. 15193025
  • 24.80 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For connecting Fast-Freeze™ flasks to freeze-dryer valves
  • 40 and 80mL flasks take size 0.5 in. or 0.5 to 0.75 in.
  • Larger flasks fit size 0.75 in. or large end of 0.5 to 0.75 in.
Specifications
Specifications
Flask Adapter
Borosilicate Glass
For connecting fast freeze™ flasks to freeze-dryer valves
0.75 in.
0.5 lb.
40, 80 mL
Straight
Fast freeze™ flask
1.9 cm
0.2 kg