missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Labconco Lid for Chamber of -50 FreeZone 4.5 L

Lids fit on drying chambers and FreeZone collector chambers.

Brand:  Labconco™ 7730700

Product Code. 11701499

  • 106.00 € / Each
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Replacement lid for 4.5L -50° C FreeZone Freeze Dryers manufactured before 2017. Verify that the part matches your FreeZone Freeze Dryer's volume and temperature rating before purchasing.

Replacement lid for 4.5 L -50° C FreeZone Freeze Dryers manufactured before 2017. Lids fit on drying chambers and FreeZone collector chambers.
Specifications

Specifications

Lid
Chamber of -50 FreeZone 4.5 L
1.1 kg
4.5 L
2.5 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.