missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Portable Table

Mobile workstation that rolls out of sight when not in use

Brand:  Labconco™ 8025000

Product Code. 13181156

  • 810.00 € / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Load capacity: 400 lb. (181kg)
  • 1 in. (2.5cm) square epoxy-coated steel frame
  • Transports heavy instruments and bulky equipment safely
  • Two open phenolic board shelves with high-pressure melamine-laminate surface resist stains and corrosion
  • 4 in. dia. (10cm) casters; two with toe locks

Order Info

Weight: 50 lb. (23kg).

Services

Lifetime guarantee.

Specifications

Specifications

Epoxy-Coated Steel Frame
Epoxy-coated
91.44 cm
88.9 cm
19 in.
35 x 19 x 36 in. (88.9 x 48.26 x 91.44 cm)
36 in.
35 in.
Steel
48.26 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.