Learn More
Labconco™ Protector XVS™ Blower Transition Adapter for Ventilation Stations
Blower Transition Adapter for Ventilation Stations
Brand:  Labconco™ 4861700
Product Code. 15479454
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • For use with Labconco Protector XVS Ventilation Stations
  • Powder-coated steel blower transition adapter allows round PVC duct to be connected to the exhaust side of the blower
Specifications
Specifications
4.25 x 7.75 in. round duct, Protector™ VS ventilation station
1.4 Kg
Epoxy-coated Steel
3 lb.
Blower Transition Adapter