Learn More
Labconco™ Protector XVS™ Zero–Pressure PVC Weathercap for Ventilation Stations
Pressure PVC Weathercap for Ventilation Stations
Brand:  Labconco™ 4722200
Product Code. 15429454
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • For use with Labconco Protector XVS Ventilation Stations
  • PVC zero-pressure weathercap connects directly to duct for vertical discharge of effluent air and dispersion away from building
  • Cap keeps out rain and snow but adds little static pressure to exhaust system
Specifications
Specifications
6 in. dia. round PVC duct for Protector™ VS ventilation station
20 lb.
PVC
36 in.
9.1 Kg