Learn More
Labconco™ Kjeldahl Replacement Parts
Replacement parts for Labconco Kjeldahl Digestion and Distillation Apparatus.
Brand:  Labconco™ 2128800
Product Code. 12753997
  • 266.00 € / Pack of 6
Explore more special offers
Description
Description
Tubes feature long necks, round perforated bottoms and thick walls to help eliminate pressure fluctuations in the distillation phase. One-piece borosilicate glass. Each tube may be label with 21288-01. Six per package. For use with Labconco Hooded Combination Kjeldahl Units, Kjeldahl Digestion Units, Kjeldahl Distillation Units, Open Combination Kjeldahl Units and RapidStill II.
Specifications
Specifications
Delivery Tube
1.4 kg
RapidStill II Unit
Borosilicate Glass
3 lb.