Learn More
Labconco™ Replacement Lid Gasket
Replacement lids for use with FreeZone 1, 2.5 & 4.5 Liter Freeze Dry Systems.
Brand:  Labconco™ 7688500
Product Code. 11856775
  • 491.00 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Replacement lids for use with FreeZone 1, 2.5 & 4.5 Liter Freeze Dry Systems.
Replacement rubber gasket allows the chamber lid of a FreeZone Freeze Dryer to seal to the chamber during the lyophilization process.
Specifications
Specifications
Lid Gasket
1, 2.5, 4.5 L
Bulk drying chamber