Learn More
Labconco™ Rotors for CentriVap™ Systems
For use with Labconco CentriVap Systems
Brand:  Labconco™ 7462900
 View more versions of this product
Product Code. 10309292
  • 382.00 € / Each
Estimated Shipment: 05-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with Labconco CentriVap Systems
  • Anodized aluminum rotors hold tubes at angle to maximize surface area
  • Two hole sizes on rotors 16-317-54 and 16-317-62 to -64 accommodate tubes with different O.D.
  • Maximum tube capacity for a given O.D. may vary with overall tube height
  • CentriVap DNA Systems and Proteomic come with DNA Rotor (16-317-54); Acid Systems come with Acid-Resistant 12-13 mm Rotor (16-108-450)
To prevent loss of sample during centrifugation, fill tubes only 0.33 to 0.5 full.
Specifications
Specifications
Anodized Aluminum
CentriVap™ DNA system
0.5 mL
0.9 kg
DNA Rotor
72 x 0.5 mL Microcentrifuge Tube or 60 x 1.5 to 2 mL Microcentrifuge Tube
2 lb.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Labconco™ Rotors for CentriVap™ Systems > DNA Rotor, Holds 72 x 0.5mL Microcentrifuge Tubes, 60 x 1.5 to 2.0mL Tubes