missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ SpillStopper™ Work Surfaces for 4ft. Protector™XStream

SpillStopper work surfaces are dished to contain spills

1915.00 €

Specifications

Material Epoxy Resin
View More Specs

Products 1
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
15963117
View Documents Promotion Details
Labconco™
9503400
No Cutouts
1915.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Front edge has large radius to aerodynamically direct airflow in the hood
  • Some surfaces have cutouts for a surface-mounted service fixture and/or a 3 x 6 in. cup sink
  • Flat work surfaces may also be coupled with an adjustable height base stand to provide auxiliary workspace
  • Service fixture and cup sink are sold separately

Specifications

Specifications

Epoxy Resin
Videos
SDS
Documents

Documents

Product Certifications