Learn More
Labconco™ Stand-Still™ Isolation Platform
The Labconco Stand-Still™ Isolation Platform is used for supporting microscopes on Purifier™ Cell Logic™+ Biosafety Cabinets with Scope-Ready™ package.
Brand:  Labconco™ 4028200
 View more versions of this product
Product Code. 15538854
  • 358.00 € / Each
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Non-porous, autoclavable Type 304 stainless steel base with 1.5 in. (0.4cm) high isolation pads
  • Reduces vertical and horizontal vibrations up to threefold, allowing proper airflow through cabinet
Stand-Still XL Platform
Requires use of Pure-Vu™ XL Seal 4027810 or 4027811 (Part Numbers)
Specifications
Specifications
12 x 18.75 in.