missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Washer Inserts, BOD Bottle Insert; Capacity: 18 Bottles

For use with Labconco FlaskScrubber™ and SteamScrubber™ Washers

Brand:  Labconco™ 4589201

Product Code. 15189675

  • 276.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

For use with Labconco FlaskScrubber™ and SteamScrubber™ Washers

Type 304 stainless-steel inserts are available in various styles to hold specific labware.

BOD Bottle Insert

  • Capacity: 18 bottles
  • Grid size: 70 × 70mm
  • With handles for easy lifting
Culture Tube Insert
  • Capacity: 242 tubes from 10 to 12mm O.D.
  • With handles for easy lifting
48-Pin Insert
  • Holds a variety of glassware
  • Stainless-steel pins are plastic-tipped to protect glassware
Petri Dish Insert
  • For Petri dishes, watch glasses, and microplates
  • Capacity: 24 plates
  • Grid size: 22 × 140mm
Pipet Insert
  • Permit direct injection pipet cleaning Capacity: 16 pipets, 1 to 50mL in size
  • Self-sealing silicone check valves allow washing of partial or full loads
  • Includes two 1.25 in. (3.2cm) mounting spindles and PVC-coated holder
  • FlaskScrubber and FlaskScrubber Vantage™ Washers hold up to two inserts
Tray Insert
  • For pans, instrument trays, multi-purpose trays and other shallow containers
  • Can be be used in lower rack of SteamScrubber or FlaskScrubber washers
  • Capacity: 5 trays

Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.