Learn More
Brady™ Thermoplastic Polyether Polyurethane Heatex™ Cable Marker
Brand:  Brady™ 622237
Product Code. 11757304
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
MIL-M-81531 AS (SAE-AS81531-1998) Point 3.4.2/4.6.2; Flame retardant; Halogen free; Resistance to solvents: MIL-STD–202G test method 215(2002) (MIL81531/SAE-AS81531-1998 Clause 3.4.3.)
Yellow
Thermoplastic Polyether Polyurethane
Heatex Cable Marker
BMP™71
250/Pk.
60.00W x 10.0 mmH
-40°C to +90°C
65.88 mm
60 mm
12.7 mm
Bulk BMP71 Heatex Cable Markers
Matte
Thermal Transfer
Voice and data communication, wire and cable marking
1
Rectangle
LabelMark
0.111 mm
10 x 60 mm
40 mm
10 mm