missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Scienceware™ Labeling Tape

Adheres to any clean surface, including Teflon™ fluoropolymer resin coated materials. Bel Art™ Labeling Tape peels off without leaving a sticky residue.

Brand:  Bel-Art™ Scienceware™ F13482-0075

29.02 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.


 View more versions of this product

Product Code. 13495226

  • 36.10 € / Pack of 4

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Excellent for use with Write-On™ Multi-Roll Label Tape Dispensers (F13472-0000 and F13473-0000)
  • Pencil, ball-point pen or solvent ink marker remain legible under the most demanding laboratory conditions
  • Consists of colored paper with rubber resin adhesive that resists moisture and most solvents
  • Will withstand a maximum temperature of 157ºC (315ºF) for 30 minutes or 80ºC (176ºF) for 24 hours; can be used down to -80ºC (-112ºF)
  • Withstands a steam autoclaving cycle at 121ºC (250ºF) and gamma irradiation between 13.0 and 45.0kGy
Specifications

Specifications

Autoclavable
Green
4/Pk.
13.7 m x 19 mm
19 mm
Self-adhesive
Label Tape
-80°C to +157°C
13.7 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Scienceware™ Labeling Tape > Color: Green; Dimensions (L x W): 13.7m x 19mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.