missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Labjack

Providing exceptionally rigid and robust support without excessive weight.

Brand:  Fisherbrand™ 2040

 View more versions of this product

Product Code. 12750027

  • 232.00 € / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Manufactured to close tolerances from aluminum with stainless steel operating screw
  • Plates have a blue anodized finish
  • Designed to be completely stable and free from rocking movements

Specifications

Specifications

Aluminum
200 x 230 mm
Max. load of 25kg
290 mm
Labjack
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.