missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Labmaster Air-Drive Mixer

Use Labmaster Air-Drive Mixers in the presence of volatile or explosive vapors, or where electrical power would be hazardous or impractical. These mixers are excellent for pilot studies of chemical, food, and wastewater treatment processes.

Brand:  Cole-Parmer™ 50322-00

 View more versions of this product

Product Code. 15457463

  • 2250.00 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • These mixers are excellent for pilot studies of chemical, food, and wastewater treatment processes.
  • LABMASTER air-drive mixers have two types of mounting styles.
    • The tube suspension mixers require the heavy-duty clamp 04560-24 (sold separately).
    • The universal clamp mixers mount directly to a ring stand or tank rim.
  • Mixers include:
    • 5/16 x 12 in., 316 SS nonthrough-type shaft
    • 5/16 x 2.5 in., 316 SS A-310 impeller
    • Collet-type chuck
    • Universal clamp or tube suspension mounting
    • Muffler
    • Needle value
  • Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications

Specifications

1800 rpm
316 Stainless Steel
37 in. oz.
Tube Suspension
1/15 hp
Air-Driven
Mixer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers