Learn More
Cytiva LabMate™ PD-10 Buffer Reservoirs
Use for high-recovery desalting, buffer exchange and removal of low-molecular weight compounds packed with Sephadex™ G-25 Medium. Cytiva Life Sciences™ LabMate™ PD-10 Buffer Reservoirs provide fast, affordable performance.
Brand:  Cytiva 18-3216-03
Product Code. 11787819
  • 55.00 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With Disposable PD-10 Desalting Columns, wash and equilibration buffer can be applied in one step with gravity flow protocol
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Buffer reservoir
10 Pack
Column Accesory
High-recovery desalting and Buffer exchange, as well as removal of low-molecular weight compounds packed with Sephadex G-25 medium, makes easier and more convenient to equilibrate PD-10 column