Learn More
Buerkle™ LaboPlast™ Bio Disposable Scoop
BIOPHARMAGREENER_CHOICE
Produced in a clean room of class 7 (10,000) and individually wrapped in bio-PE film for disposable use. Green PE bio-plastic has all the suitable properties of conventional PE but is manufactured from renewable raw materials.
Brand:  Buerkle™ 5379-0005
24.23 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15386327
  • 28.25 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled
  • The scoop stands horizontally as a result of the special handle design and the slightly angled front edge and can therefore be put down without the sample material trickling out.
  • Product and packaging made of renewable raw materials
  • Bio-PE, white
  • Individually wrapped
  • Production and packaging according to clean room class 7
  • With EU foodstuffs and FDA approval
Specifications
Specifications
Polyethylene
Single-use
White
Foodstuffs
205 mm
100 mL
Non-sterile
FDA
10