Learn More
Buerkle™ LaboPlast™ Disposable Scoop
BIOPHARMAGREENER_CHOICE
The LaboPlast™ disposable scoop made of polystyrene is produced and assembled in a clean room of class 7 and individually packaged for disposable use. The disposable scoop is ideal for powders, granulates and pastes.
Brand:  Buerkle™ 5378-0006
32.96 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15396307
  • 37.40 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Scoop, disposable with or without cover
  • Polystyrene, white
  • Individually packaged
  • Production and packaging according to Cleanroom Class 7
  • With EU Foodstuffs and FDA approval
Specifications
Specifications
Disposable Scoop
With Cover
100 mL
206 mm
Polystyrene
Cover
Single-use