missing translation for 'onlineSavingsMsg'
Learn More

Lancer™ Laboratory Machine Detergent

Use in both Lancer and other quality washing machines. Lancer™ Laboratory Machine Detergent has been specially formulated to give the best possible results.

225.27 € - 241.00 €

Specifications

Product Type Laboratory Machine Detergent
For Use With (Application) Formulated to give the best possible results in both Lancer and other quality washing
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
13112230
View Documents Promotion Details
Lancer™
41010070
10 L
241.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
13122230
View Documents Promotion Details
Lancer™
LCD25
25 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Chemical cleaning compound
  • Health and Safety Data Sheets available to comply with CHP regulations
Specifications

Specifications

Laboratory Machine Detergent
Formulated to give the best possible results in both Lancer and other quality washing
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Carcinogenicity Category 2

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P264-Wash thoroughly after handling.
  • P405-Store locked up.

Supplemental information
  • MIXTURE LIST-Contains : Trisodium nitrilotriacetate, Sodium hydroxide

SDS
Documents

Documents

Product Certifications