Learn More
Cytiva (formerly Pall Lab) Laboratory Manifold 3/8 in. Hose Barb End Cap
Replacement tubing outlet accessory for microbiology manifold.
Brand:  Cytiva (formerly Pall Lab) 4967
Product Code. 17184451
Description
Description
  • Lab manifold is designed for flexibility and reduction in cross-contamination
  • Lab manifold features interchangeable adapters for most funnels, and a coupling device to double testing capacity while still enabling sterilization in standard autoclaves
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Laboratory Manifold
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva (formerly Pall Lab) Laboratory Manifold 3/8 in. Hose Barb End Cap > For use with Laboratory Manifold