Learn More
Epredia™ Laboratory Marking Pen, Secureline Marker II
Product Code. p-6021136 Shop All Epredia Products
Click to view available options
Product Type:
Laboratory Marking Pen
Secureline Marker II
Unit Size:
Each
Pack of 10
Product Code. 15582321
Brand: Epredia™ 2000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Mark cassettes and microscope slides with the Epredia™ Laboratory Marking Pen.
  • Resistant to common laboratory solvents (water, 10% neutral buffered formalin, alcohol)
  • Extra fine point for precise writing
  • Non-sterile
  • Not made with natural rubber latex
TRUSTED_SUSTAINABILITY
Specifications
Product Type Laboratory Marking Pen
Color Black
Quantity 10/Pk.