missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Electro™ Nickel Crucibles
Get optimum results with nickel crucibles. With working temperature ranges up to 350°C, the Nickel Electro™ Nickel Crucible Capacity consistently has a long working life.
Brand: Nickel Electro™ 40
This item is not returnable.
View return policy
Description
- Tall form, without lid
- Suitable for strong alkalis and phosphoric acid
- Resistant to HF and Cl2below 500°C and to HCl
- Capacity and dimensions are approximate
- Working temperature range up to 350°C will ensure crucible has a long working life
- In the analytical laboratory, nickel crucibles provide a low cost crucible
- Can be used for research, experiments, chemical analysis, tests, quality control and education
Specifications
Nickel | |
Each | |
130 mL | |
60 mm |
350°C | |
2 mm | |
60 mm | |
Laboratory Crucible |