missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Laboratory Notebooks
GREENER_CHOICE

Record research data, experimental procedures, and design ideas. Laboratory Notebooks contain periodic and conversion tables in the back.

Brand:  Heathrow Scientific™ HS8610D

1883.61 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 11304045

  • 2630.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Numbered, acid-free, pages
  • Include spaces for title, project number, book number, date, and witness signature
  • Lined notebooks feature college-ruled pages for easy report writing
  • Covers for all versions are waterproof and chemical-resistant
  • Measures 218 x 281 mm
Specifications

Specifications

Acid-free Paper
8.5 in.
Red
11 x 8.5 in. (281 x 218 mm)
21.6 cm
Laboratory Notebook
Yes
11 in.
Red
27.9 cm
200
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.