missing translation for 'onlineSavingsMsg'
Learn More

Laboratory Buffer Solution, pH 9.21, Blue, Reagecon™
BIOPHARMA

Product Code. 11796388 Shop All Reagecon Products
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
This item is not returnable. View return policy

Product Code. 11796388

Brand: Reagecon 90515

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for use in laboratory pH measurement. Reagecon™ Laboratory Buffer Solution, pH 9.21, Blue is tested at 20°C to an accuracy level of ±0.02. This calibration and control buffer is specially formulated to ensure its stability and is supplied with a detailed Certificate of Analysis.

Reagecon manufactures the most comprehensive range of pH reagents in the world; these are designed to suit all end user requirements. This laboratory grade buffer is manufactured to exacting specifications with an extended shelf life and is supplied with a detailed Certificate of Analysis and comprehensive label. It is color-coded for easy identification.

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Blue
pH 9.21 ±0.02
Physical Form Liquid
Quantity 250 mL
Odor Characteristic
Solubility Information Fully miscible in water
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.