Learn More
Welch Ilmvac™ Labovac 10 Oil
Use this mineral oil with standard rotary vane vacuum pumps. Welch Ilmvac™ Labovac 10 Oil is ideal for a variety of applications.
Brand:  Welch Ilmvac™ 800123
Product Code. 11819580
  • 276.00 € / 10 L
Explore more special offers
Description
Description
  • Mineral oil based vacuum oil for standard rotary vane vacuum pumps
  • 10L
  • For applications such as pumping out air, inert gases and water vapor, and also slightly acidic or basic vapors
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vacuum Oil
10 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Welch Ilmvac™ Labovac 10 Oil > Volume: 10L