Learn More
Thermo Scientific™ Labtainer™ 2D BioProcess Container, BioTitan retainers, two ports, Luer locks
The Thermo Scientific 2D Labtainer with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.
Brand: Thermo Scientific™ SH31292.14
Description
The Thermo Scientific 2D Labtainer with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.
Features of the Thermo Scientific 2D BPC system include:
• Now manufactured with the BioTitan Retention Device—designed to provide superior universal retention that minimizes the risk of leakage and failure at the connection point
• Use to eliminate post-use cleaning steps required with re-useable containers and to reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions
Applications of the Thermo Scientific 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Buffer, culture media, and process liquid preparation and storage
• Chromatography feed and fraction collection
• Harvest, storage, and transport of bulk drug product and bulk drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
Specifications
500 mL | |
TPE |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.